Learn More
Cayman Chemical Shepherdin (trifluoroacetate salt), Purity: ≥90%, 10 mg
SDP
Catalog No. 502918027
Encompass_Preferred
An inhibitor of the survivin-Hsp90 interaction; binds to the N-terminal domain of Hsp90; inhibits the interaction between recombinant human survivin and Hsp90 in isolated human reticulocyte extracts at 150 µM; peptide conjugates with penetratin or HIV Tat sequence have anticancer activity in vitro and in vivo
Catalog No. 50-291-8027 Supplier Cayman Chemical Supplier No. 3832710 MG
An inhibitor of the survivin-Hsp90 interaction; binds to the N-terminal domain of Hsp90; inhibits the interaction between recombinant human survivin and Hsp90 in isolated human reticulocyte extracts at 150 µM; peptide conjugates with penetratin or HIV Tat sequence have anticancer activity in vitro and in vivo