Learn More
ATCC Lactiplantibacillus pentosus
Supplier:  ATCC 8041
Encompass
Lactiplantibacillus pentosus strain 124-2 is a whole-genome sequenced bacterial type strain that has applications in lactic acid production, acetic acid production, sawdust degradation, pentose fermentation, and hexose fermentation.
Catalog No. 50-237-9704