Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ShhN (SHH) ELISA Kit

Catalog No. EHSHHX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHSHHX5 5 x 96 Tests
EHSHH 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHSHHX5 Supplier Invitrogen™ Supplier No. EHSHHX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Human ShhN (SHH) ELISA quantitates Hu SHH in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu SHH. Principle of the method The Human SHH solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Sonic Hedgehog (SHH), which is expressed only during embryogenesis, is instrumental in patterning the early embryo. It has been implicated as the key inductive signal in patterning of the ventral neural tube, the anterior-posterior limb axis, and the ventral somites. Of three human proteins showing sequence and functional similarity to the Sonic Hedgehog protein of Drosophila, this protein is the most similar. The protein is made as a precursor that is autocatalytically cleaved, the N-terminal portion is soluble and contains the signalling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the developing embryo. Defects in this protein or in its signalling pathway are a cause of holoprosencephaly (HPE), a disorder in which the developing forebrain fails to correctly separate into right and left hemispheres. HPE is manifested by facial deformities. In addition, it is thought that mutations in this gene or in its signalling pathway may be responsible for VACTERL syndrome, which is characterized by vertebral defects, anal atresia, tracheoesophageal fistula with esophageal atresia, radial and renal dysplasia, cardiac anomalies, and limb abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15465
Assay Range 8-10000 pg/mL
Assay Sensitivity 8 pg/mL
Calibration Range 8 pg/mL to 10,000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Proteases Peptidases & Inhibitors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Sufficient For 5 x 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 9530036O11Rik; Dsh; fc83d08; hedgehog; hemimelic extra toes; Hhg1; HHG-1; HLP3; HPE3; Hx; Hxl3; M100081; MCOPCB5; morphogen; shh; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; shh.L; shha; ShhN; ShhNC; ShhNp; short digits; SMMCI; sonic hedgehog; sonic hedgehog a; sonic hedgehog homolog; sonic hedgehog L homeolog; sonic hedgehog protein; Sonic hedgehog protein 19 kDa product; Sonic hedgehog protein A; Sonic hedgehog protein A N-product; Sonic hedgehog protein N-product; sonic hedgehog signaling molecule; sonic hedgehog signaling molecule a; sonic-you; syu; TPT; TPTPS; vhh1; vhh-1; wu:fc83d08; XELAEV_18030676mg; Xhh; xshh; X-SHH9530036O11Rik; Dsh; fc83d08; hedgehog; hemimelic extra toes; Hhg1; HHG-1; HLP3; HPE3; Hx; Hxl3; M100081; MCOPCB5; morphogen; shh; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; shh.L; shha; ShhN; ShhNC; ShhNp; short digits; SMMCI; sonic hedgehog; sonic hedgehog a; sonic hedgehog homolog; sonic hedgehog L homeolog; sonic hedgehog protein; Sonic hedgehog protein 19 kDa product; Sonic hedgehog protein A; Sonic hedgehog protein A N-product; Sonic hedgehog protein N-product; sonic hedgehog signaling molecule; sonic hedgehog signaling molecule a; sonic-you; syu; TPT; TPTPS; vhh1; vhh-1; wu:fc83d08; XELAEV_18030676mg; Xhh; xshh; X-SHH; HHG1; HLP3; HPE3; MCOPCB5; SMMCI; TPT; TPTPS
Gene ID (Entrez) 6469
Gene Symbol Shh
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases SHH, HHG-1
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Development
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym tcag7.582, HHG1, HLP3, HPE3, MCOPCB5, SMMCI, TPT, TPTPS
Target SHH
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.