Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ SHIBAM Agar
SDP

Catalog No. 23001811 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Species:
Escherichia coli
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-001-811 Escherichia coli
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-811 Supplier Hardy Diagnostics™ Supplier No. A146
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a selective and differential growth medium for the cultivation of Shiga toxin-producing E. coli.

  • Detects enterohemolysin producing E. coli
  • Zones of clear blood hemolysis (beta hemolysis) surrounding the colonies are associated with Shiga-toxin producing E. coli
  • Formulation recommended by BAM

Specifications

Color Red
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Mitomycin C, Washed Defibrinated Sheep Blood
Detects Shiga Toxin
Format Plate
Product Type Agar Plate
Sterility Sterile
Target Organism Class E. coli
Test Type Microbial Detection
Fill Volume 17 mL
Formulation Agar - 14.0 gm, Peptones - 17.0 gm, Sodium Chloride - 5.0 gm, Tryptose - 5.0 gm, Yeast Extract - 4.0 gm, Calcium Chloride - 1.47 gm, Mitomycin C - 0.5 mg, Washed Defibrinated Sheep Blood - 40 ml per liter of deionized water
For Use With (Application) Cultivation of Shiga toxin-producing E. coli (STEC)
Incubation Conditions 35°C to 37°C, Aerobically
Incubation Time 16 to 24 hr.
Packaging Type Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Escherichia coli
Protein Source Peptones, Tryptose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.