Learn More
Selleck Chemical LLC Shield-1 5mg
SDP
Supplier:  Selleck Chemical LLC S34695MG
Encompass_Preferred
Shield-1 is a specific, cell-permeant and high-affinity ligand of FK506-binding protein-12 (FKBP). Shield-1 reverses the instability by binding to mutated FKBP (mtFKBP), allowing conditional expression of mtFKBP-fused proteins. Shield-1 stabilizes the entire fusion protein. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3116