Learn More
TARGETMOL CHEMICALS INC SHIELD-1 5MG
SDP
Catalog No. 502581322
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Shield-1 is a specific high-affinity and cell-permeant ligand of FK506-binding protein-12 (FKBP) and reverses the instability by binding to mutated FKBP (mtFKBP) allowing conditional expression of mtFKBP-fused proteins. Shield-1 can stabilize the entire fusion protein. purity: 99%
Catalog No. 50-258-1322 Supplier TARGETMOL CHEMICALS INC Supplier No. T138845MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Shield-1 is a specific high-affinity and cell-permeant ligand of FK506-binding protein-12 (FKBP) and reverses the instability by binding to mutated FKBP (mtFKBP) allowing conditional expression of mtFKBP-fused proteins. Shield-1 can stabilize the entire fusion protein. purity: 99%