Learn More
MSC Zep Floor Polish, Slip-Resistant, High Durability, Acrylic, 55 Gal
Catalog No. 502867665
Encompass
Shield Floor Polish is a milky white, slip-resistant, water-based emulsion and acrylic copolymer. It offers high durability and low maintenance, with a metal-interlocked formula resistant to traffic scuffs and black heel marks. This dry-bright polish is easy to apply, levels without streaks, and blends well with existing polish.
  • High durability; metal-interlocked to resist traffic scuffs and black heel marks
  • Each coat dries in 30 minutes
  • Easy to apply and level; recoats and blends well with existing polish
  • Low maintenance; gloss not affected by neutral detergents
  • Meets or exceeds slip resistance standards established by CSPA and UL as tested in accordance with ASTM D-2047
  • Easily removed by any floor stripper
  • Can be spray-buffed with Buff-Up and a red pad with a low-speed floor machine
  • Milky white color
Catalog No. 50-286-7665 Supplier MSC Supplier No. 17690249
Shield Floor Polish is a milky white, slip-resistant, water-based emulsion and acrylic copolymer. It offers high durability and low maintenance, with a metal-interlocked formula resistant to traffic scuffs and black heel marks. This dry-bright polish is easy to apply, levels without streaks, and blends well with existing polish.
  • High durability; metal-interlocked to resist traffic scuffs and black heel marks
  • Each coat dries in 30 minutes
  • Easy to apply and level; recoats and blends well with existing polish
  • Low maintenance; gloss not affected by neutral detergents
  • Meets or exceeds slip resistance standards established by CSPA and UL as tested in accordance with ASTM D-2047
  • Easily removed by any floor stripper
  • Can be spray-buffed with Buff-Up and a red pad with a low-speed floor machine
  • Milky white color