Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ SHIELD™ Floor and Wall Protector for FMX™ Dispensers
SDP

Catalog No. 19043912 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Mounting:
Wall Mount
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
19-043-912 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-043-912 Supplier GOJO™ Supplier No. 276106
  • $64.00
    $47.65 / Case of 12

    Save $16.35

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protects floors and walls against splashes and drips caused by typical use.

  • Helps protect floors, walls and countertops
  • Attaches to any FMX™ dispenser
  • Easy to install
TRUSTED_SUSTAINABILITY

Specifications

Product Type Floor and Wall Protector
Color Gray
Length (Metric) 3.88 cm
Width (English) 4.56 in.
Width (Metric) 4.56 cm
Height (English) 6.31 in.
Height (Metric) 6.31 cm
Holds Hand Sanitizer
Material Plastic
Mounting Wall Mount
No. of Compartments 1
Product Style Front-loading with Opening
For Use With (Equipment) FMX™ Dispensers
Quantity 6/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.