Learn More
Med Vet International Animal Shield Large, 24"x24", J1392A, Each
Catalog No. 5000495947
Encompass_Preferred
Just slip this light transparent panel into the cage, push toward the rear, and inject through the evenly spaced holes.– Easy to clean– Safe for the handler– Humane to the animal– Integral handle for one handed grip24? x 24? to fit larger cages
Catalog No. 50-004-95947 Supplier Med Vet International Supplier No. JOR1392A
Just slip this light transparent panel into the cage, push toward the rear, and inject through the evenly spaced holes.– Easy to clean– Safe for the handler– Humane to the animal– Integral handle for one handed grip24? x 24? to fit larger cages