Learn More
LIGHTHOUSE WORLDWIDE SOLUTIONS INC SHIELD PRO N95 4X20CS 80CS
SDP
Catalog No. NC3943218
Encompass
Designed For Superior Fit The SHIELD-PRO is designed to fit a wide range of facial features, but on top of that, its unique build allows for maximum flexibility. Even as your face moves and you make expressions, the SHIELD-PRO is designed to keep its seal. You shouldn’t have to change your mannerisms out of concern for your safety and to cater to a respirator, which is why our SHIELD-PRO works hard for you.Reaching New Levels of Breathability Not only should your respirator stay sealed, it should be comfortable. Our unique 3-panel design allows for not only a larger filtration surface, but improved breathability. In fact, the SHIELD-PRO is 15% more breathable than competitors’ options. Maximizing VisibilityYou work in dangerous situations and you need your vision. Annoying fog, limited field of vision, and general discomfort can make respirators a bother. But LWS’ SHIELD-PRO is designed to provide 25% more visibility with our
Catalog No. NC3943218 Supplier LIGHTHOUSE WORLDWIDE SOLUTIONS INC Supplier No. 900MISCPART/CS
Designed For Superior Fit The SHIELD-PRO is designed to fit a wide range of facial features, but on top of that, its unique build allows for maximum flexibility. Even as your face moves and you make expressions, the SHIELD-PRO is designed to keep its seal. You shouldn’t have to change your mannerisms out of concern for your safety and to cater to a respirator, which is why our SHIELD-PRO works hard for you.Reaching New Levels of Breathability Not only should your respirator stay sealed, it should be comfortable. Our unique 3-panel design allows for not only a larger filtration surface, but improved breathability. In fact, the SHIELD-PRO is 15% more breathable than competitors’ options. Maximizing VisibilityYou work in dangerous situations and you need your vision. Annoying fog, limited field of vision, and general discomfort can make respirators a bother. But LWS’ SHIELD-PRO is designed to provide 25% more visibility with our