Learn More
Med Vet International Small Animal Shield, for Cats, J1392
Catalog No. 5000495804
Encompass_Preferred
Just slip this light transparent panel into the cage, push toward the rear, and inject through the evenly spaced holes.– Easy to clean– Safe for the handler– Humane to the animal– Integral handle for one handed grip15? x 15? to fit 23?x 23? cage
Catalog No. 50-004-95804 Supplier Med Vet International Supplier No. JOR1392
Just slip this light transparent panel into the cage, push toward the rear, and inject through the evenly spaced holes.– Easy to clean– Safe for the handler– Humane to the animal– Integral handle for one handed grip15? x 15? to fit 23?x 23? cage