Learn More
Takara Bio Shield1, 500 ul
SDP
Catalog No. NC0159458
Encompass
The Shield1 compound stabilizes a protein of interest tagged with the destabilization domain (DD), which is based on a mutated version of the FKBP protein. It is used to protect the DD fusion protein of interest from proteasomal degradation. The amount of Shield1 can be varied to tune the amount of stabilized protein of interest within the cell. 500 ul
Catalog No. NC0159458 Supplier Takara Bio Supplier No. 632189
The Shield1 compound stabilizes a protein of interest tagged with the destabilization domain (DD), which is based on a mutated version of the FKBP protein. It is used to protect the DD fusion protein of interest from proteasomal degradation. The amount of Shield1 can be varied to tune the amount of stabilized protein of interest within the cell. 500 ul