Learn More
Bioss Shiga toxin
SDP
Catalog No. 5000065737
Encompass_Preferred
Shiga toxin Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65737 Supplier Bioss Supplier No. BSM72416M
Shiga toxin Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide