Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vela Diagnostics Great Basin Shiga Toxin Direct Test

Catalog No. 23052004
Change view
Click to view available options
Quantity:
10 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-052-004 10 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-052-004 Supplier Vela Diagnostics Supplier No. 300783
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A sample-to-result, automated, assay that detects Shiga toxin-producing E. coli directly from patient’s stool sample in <2 hours. The assay detects stx1 and stx2, in addition to identifying the hypervirulent serotype O157. Unlike competing tests on the market, this assay does not require an enrichment step.

Utilizes a fully enclosed, disposable cartridge with all the reagents needed to detect the stx1 and stx2 genes, and additional E. coli O157 identification in Shiga toxin positive specimens, without culture for an accurate and efficient patient result.

Specifications

Content And Storage 2°C to 8°C
Description Sample-to-result test: Shiga toxin
Clia Complexity Moderate
Detectable Analytes E. coli Shiga Toxin 1 and 2, E. coli Serogroup O157, E. coli
Sample Type Preserved Stool
Quantity 10 Assays
Sample Size 250 μL
Test Time ≈90 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.