Learn More
ATCC Shigella flexneri 2a; Strain 2457T
Catalog No. 502380664
Encompass
Shigella flexneri strain 2457T is a whole-genome sequenced bacterium with applications in enteric disease research. The presence of the plasmid pINV-2457T has not been confirmed.
Catalog No. 50-238-0664 Supplier ATCC Supplier No. 700930
Shigella flexneri strain 2457T is a whole-genome sequenced bacterium with applications in enteric disease research. The presence of the plasmid pINV-2457T has not been confirmed.