Learn More
Bioss Shigella sonnei form 1
SDP
Catalog No. 5000065366
Encompass_Preferred
Shigella sonnei form 1 Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65366 Supplier Bioss Supplier No. BSM72425M
Shigella sonnei form 1 Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide