Learn More
ATCC Shigella sonnei; Strain NCDC 1120-66
Catalog No. 502383802
Encompass
Shigella sonnei strain NCDC 1120-66 [CIP 104223] was isolated from human feces. This whole-genome sequenced bacterium is a quality control strain for bioMerieux VITEK and Difco products.
Catalog No. 50-238-3802 Supplier ATCC Supplier No. 25931
Shigella sonnei strain NCDC 1120-66 [CIP 104223] was isolated from human feces. This whole-genome sequenced bacterium is a quality control strain for bioMerieux VITEK and Difco products.