Learn More
Sigma Aldrich Fine Chemicals Biosciences Shigemi 5 mm Symmetrical N1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5293461EA
Encompass
Shigemi NMR microtubes offer high precision quality in NMR spectroscopy. The NMR microtubes are made out of a special type of hard glass wtih excellent chemical durability and are magnetic susceptibility matched to each of the solvents they are identified with.The Symmetrical NMR microtube consists of outer tube and insert with these featuresMagnetic susceptibility matched to a variety of solvents for the best reSOLUTIONon in these solvents using minimal amount of sampleThe small sample size will not affect the spectral reSOLUTIONon or the sidebandsEach microtube assembly consists of an insert and an outer sample tubeSuitable for 900 MHz NMR instrumentsChoose a bottom length that most closely matches the probe coil.
Catalog No. 50-290-1218