Learn More
Medchemexpress LLC HY-N0130 100mg Medchemexpress, Shikimic acid CAS:138-59-0 Purity:>98%
SDP
Catalog No. 501933412
Encompass_Preferred
Medchemexpress, HY-N0130 100mg Shikimic acid CAS:138-59-0 Shikimic acid is a key metabolic intermediate of the aromatic amino acid biosynthesis pathway, found in microbes and plants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3412 Supplier Medchemexpress LLC Supplier No. HYN0130100MG
Medchemexpress, HY-N0130 100mg Shikimic acid CAS:138-59-0 Shikimic acid is a key metabolic intermediate of the aromatic amino acid biosynthesis pathway, found in microbes and plants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.