Learn More
Sigma Aldrich Fine Chemicals Biosciences (2R,3R,4S)-3-(Hydroxymethyl)-3,4,5,6-tetrahydroxy-tetrahydro-2H-pyran-2-carboxylic Acid | 138-59-0 | 25 GR
Catalog No. NC2670412
Encompass
Shikimic acid | Purity: ≥99% | Mol Wt: 174.15 | CAS No: 138-59-0 | MFCD00001392 | 25gThis organic compound serves as a chiral starting material for synthesizing aromatic amino acids like phenylalanine, tyrosine, and tryptophan through the shikimic acid pathway. It contains functional groups including a carboxylic acid and three hydroxyls.
Catalog No. NC2670412 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S537510MG
Shikimic acid | Purity: ≥99% | Mol Wt: 174.15 | CAS No: 138-59-0 | MFCD00001392 | 25gThis organic compound serves as a chiral starting material for synthesizing aromatic amino acids like phenylalanine, tyrosine, and tryptophan through the shikimic acid pathway. It contains functional groups including a carboxylic acid and three hydroxyls.