Learn More
Medchemexpress LLC Shikimic acid | 138-59-0 | 96.9% | 174.15 | 25 MG
SDP
Catalog No. 5000430479
Encompass_Preferred
Shikimic acid is a key metabolic intermediate within the aromatic amino acid biosynthesis pathway, present in microbes and plants. It serves as an industrially significant compound and a precursor in the synthesis of various chemical substances. The product is a white to off-white solid, supplied in a brown glass bottle, and is suitable for use as an analytical standard.
  • Key metabolic intermediate in aromatic amino acid biosynthesis
  • Precursor in the synthesis of various chemical substances
  • Supplied as a white to off-white solid
  • Packaged in a brown glass bottle for optimal storage
  • Suitable for use as an analytical standard
  • Fully qualified and calibrated to ISO 17025 requirements
  • Organization certified to ISO 9001 and ISO/IEC 17025
Catalog No. 50-004-30479 Supplier Medchemexpress LLC Supplier No. 25MGHYN0130R
Add to Cart
Add to Cart
Shikimic acid is a key metabolic intermediate within the aromatic amino acid biosynthesis pathway, present in microbes and plants. It serves as an industrially significant compound and a precursor in the synthesis of various chemical substances. The product is a white to off-white solid, supplied in a brown glass bottle, and is suitable for use as an analytical standard.
  • Key metabolic intermediate in aromatic amino acid biosynthesis
  • Precursor in the synthesis of various chemical substances
  • Supplied as a white to off-white solid
  • Packaged in a brown glass bottle for optimal storage
  • Suitable for use as an analytical standard
  • Fully qualified and calibrated to ISO 17025 requirements
  • Organization certified to ISO 9001 and ISO/IEC 17025