Learn More
Cayman Chemical Shikimic Acid, Purity: ≥95%, CAS: 138-59-0, 500 mg
SDP
Catalog No. 502934788
Encompass_Preferred
A cyclohexenecarboxylic acid; an intermediate in the biosynthesis of aromatic amino acids in plants and microorganisms; a precursor in the synthesis of oseltamivir; decreases lipid droplet accumulation in HepG2 cells and 3T3-L1 adipocytes at 80 µM; increases the levels of phosphorylated AMPK and decreases the levels of MID1IP1 in HepG2 and 3T3-L1 adipocytes at 40 µM; decreases inflammation and demyelination in a mouse model of EAE at 50-200 mg/kg and reduces symptom progression at 100 mg/kg
Catalog No. 50-293-4788 Supplier Cayman Chemical Supplier No. 26851500 MG
A cyclohexenecarboxylic acid; an intermediate in the biosynthesis of aromatic amino acids in plants and microorganisms; a precursor in the synthesis of oseltamivir; decreases lipid droplet accumulation in HepG2 cells and 3T3-L1 adipocytes at 80 µM; increases the levels of phosphorylated AMPK and decreases the levels of MID1IP1 in HepG2 and 3T3-L1 adipocytes at 40 µM; decreases inflammation and demyelination in a mouse model of EAE at 50-200 mg/kg and reduces symptom progression at 100 mg/kg