Learn More
Apexbio Technology LLC SHIKIMIC-ACID-50MG
SDP
Catalog No. 5000569273
Encompass_Preferred
Shikimic acid (CAS No 138-59-0) is primarily investigated in the fields of biochemistry and pharmaceuticals originating from the plant Illicium verum commonly known as star anise It plays a critical role in the shikimate pathway which is pivotal in the biosynthesis of aromatic compounds in microorganisms and plants This pathway is essential for the production of folates and aromatic amino acids making shikimic acid a significant subject of study In vitro shikimic acid demonstrates activity in the nanomolar to low micromolar range highlighting its potential influence on biological processes It is frequently used in cell models and animal studies to explore its potential as a precursor in drug development particularly in the synthesis of oseltamivir an antiviral medication Shikimic acid is also valuable in drug screening assays for evaluating biochemical pathways and mechanisms of action Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69273 Supplier Apexbio Technology LLC Supplier No. N182950
Add to Cart
Add to Cart
Shikimic acid (CAS No 138-59-0) is primarily investigated in the fields of biochemistry and pharmaceuticals originating from the plant Illicium verum commonly known as star anise It plays a critical role in the shikimate pathway which is pivotal in the biosynthesis of aromatic compounds in microorganisms and plants This pathway is essential for the production of folates and aromatic amino acids making shikimic acid a significant subject of study In vitro shikimic acid demonstrates activity in the nanomolar to low micromolar range highlighting its potential influence on biological processes It is frequently used in cell models and animal studies to explore its potential as a precursor in drug development particularly in the synthesis of oseltamivir an antiviral medication Shikimic acid is also valuable in drug screening assays for evaluating biochemical pathways and mechanisms of action Experimental concentrations typically depend on the specific research design