Learn More
Medchemexpress LLC HY-N0822 10mg , Shikonin CAS:517-89-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN082210MG
Encompass_Preferred
HY-N0822 10mg Shikonin CAS: 517-89-5 Shikonin is a major component of a Chinese herbal medicine named zicao. Shikonin is a potent TMEM16A chloride channel inhibitor with an IC50 of 6.5 μM. Shikonin is a specific pyruvate kinase M2 (PKM2) inhibitor and can also inhibit TNF-α and NF-κB pathway. Shikonin decreases exosome secretion through the inhibition of glycolysis[4]. Shikonin inhibits AIM2 inflammasome activation[7]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1179