Learn More
Selleck Chemical LLC Shikonin 20mg 54952-43-1 Anchusin, Anchusa acid, Alkanna Red, C.I. 75535, Isoarnebin 4, NSC 252844
SDP
Supplier:  Selleck Chemical LLC S827920MG
Encompass_Preferred
Shikonin (nchusin, Anchusa acid, Alkanna Red, C.I. 75535, Isoarnebin 4, NSC 252844), a potent and specific Pyruvate kinase M2 (PKM2) inhibitor, is a major component of zicao (purple gromwell, the dried root of Lithospermum erythrorhizon), a Chinese herbal medicine with various biological activities. It is also an inhibitor of TMEM16A chloride channel activity using cell-based fluorescent-quenching assay. Shikonin exerts an anti-inflammatory effect by inhibiting tumor necrosis factor-? (TNF-?) and prevents activation of nuclear factor-?B (NF-?B) pathway via proteasome inhibition. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5888