Learn More
Medchemexpress LLC HY-N0822 100mg , Shikonin C.I. 75535;Isoarnebin 4 CAS:517-89-5 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYN0822100MG
Encompass_Preferred
Medchemexpress, HY-N0822 100mg Shikonin C.I. 75535;Isoarnebin 4 CAS:517-89-5 Formula:C16H16O5 IC50: 6.5 μM (TMEM16A chloride channel) PKM2 TNF-α, NF-κB Purity:98% Shikonin is an inhibitor of TMEM16A chloride channel with an IC 50 of 6.5 μM. Shikonin is also a specific inhibitor of PKM2 and can also inhibit tumor necrosis factor-α ( TNF-α ) and prevent activation of nuclear factor-κB ( NF-κB ) pathway. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2935
 Restricted Item