Learn More
AdipoGen Shikonin (50 mg)
SDP
Supplier:  AdipoGen AGCN20487M050
Encompass_Preferred
Chemical. CAS: 517-89-5. Formula: C16H16O5. MW: 288.3. Isolated from Alkanna sp. Anticancer compound. Inhibits TNF-alpha-induced and B-16 melanoma-induced angiogenesis. Induces apoptosis and RIP1- and RIP3-dependent necroptosis in several cancer cells. Circumvents cancer antidrug resistance through the necroptosis pathway. Proteasome inhibitor. Autophagy inducer. Topoisomerase I inhibitor. Inhibits glycolysis in cancer cells by inhibiting tumor-specific pyruvate kinase M2 (PKM2). Anti-inflammatory compound. Inhibits leukocyte migration, downregulates chemokine receptor expression, and inhibits HIV-1 replication. Inhibits the activation of NLRP3 and AIM2 inflammasome. Shown to directly target caspase-1 and as a inhibitor of PKM2 to block PKM2-mediated glycolysis that promotes inflammasome activation by modulating EIF2AK2 phosphorylation in macrophages. Antioxidant. Free radical scavenger. Directly inhibits nitric oxide synthase (NOS).
Catalog No. 50-205-3671