Learn More
Glass Expansion Shimadzu ICP-MS 2030, Gold Coil. To ensure correct installation, the Installation Kit 70-900-6000T is required. This is not included and must be ordered separately. It is re-useable so only needs to be ordered once.
SDP
Catalog No. 501907337
Encompass_Preferred
RF Coil Gold for Shimadzu 2030 ICP-MS. Compatible with Shimadzu 2030 ICP-MS. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. For Copper 501907336, Silver 501907338.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit (501907339).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7337 Supplier Glass Expansion Supplier No. 709006000G
RF Coil Gold for Shimadzu 2030 ICP-MS. Compatible with Shimadzu 2030 ICP-MS. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. For Copper 501907336, Silver 501907338.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit (501907339).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001