Learn More
Glass Expansion Shimadzu ICPE 9800 Installation Tool
SDP
Catalog No. 501907343
Encompass_Preferred
RF Coil Installation Tool for Shimadzu ICPE 9800 ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for Shimadzu ICPE 9800 ICP-OES, P/Ns 501907340, 501907341 and 501907342. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7343 Supplier Glass Expansion Supplier No. 709006100T
RF Coil Installation Tool for Shimadzu ICPE 9800 ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for Shimadzu ICPE 9800 ICP-OES, P/Ns 501907340, 501907341 and 501907342. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001