Learn More
Analytical Sales & Services Shimadzu Model LC-10AT Self-Pr
Catalog No. 5000806748
Encompass
Shimadzu Model LC-10AT Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM No. 228-32166-91\n\nQty/Pkg: EACH
Catalog No. 50-008-06748 Supplier Analytical Sales & Services Supplier No. 925184
Shimadzu Model LC-10AT Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM No. 228-32166-91\n\nQty/Pkg: EACH