Learn More
Analytical Sales & Services Shimadzu Models LC-10ADvp & LC
Catalog No. 5000804268
Encompass
Shimadzu Models LC-10ADvp & LC-20AD & ESA Models 580 & 582 Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM No. 228-39093-92 & 228-45704-91\n\nQty/Pkg: EACH
Catalog No. 50-008-04268 Supplier Analytical Sales & Services Supplier No. 925181
Shimadzu Models LC-10ADvp & LC-20AD & ESA Models 580 & 582 Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM No. 228-39093-92 & 228-45704-91\n\nQty/Pkg: EACH