Learn More
Analytical Sales & Services Shimadzu Models LC-10ATvp LC-
Catalog No. 5000806785
Encompass
Shimadzu Models LC-10ATvp LC-2010 & LC-20AT Self-Priming Outlet Check Valve Cartridge Only
Qty/Pkg: EACH
Catalog No. 50-008-06785 Supplier Analytical Sales & Services Supplier No. 925168
Shimadzu Models LC-10ATvp LC-2010 & LC-20AT Self-Priming Outlet Check Valve Cartridge Only
Qty/Pkg: EACH