Learn More
Analytical Sales & Services Shimadzu Models LC-10ATvp LC-
Catalog No. 5000804267
Encompass
Shimadzu Models LC-10ATvp LC-2010 & LC-20AT Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM No. 28-32166-91
Qty/Pkg: EACH
Catalog No. 50-008-04267 Supplier Analytical Sales & Services Supplier No. 925140
Shimadzu Models LC-10ATvp LC-2010 & LC-20AT Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM No. 28-32166-91
Qty/Pkg: EACH