Learn More
Analytical Sales & Services Shimadzu Models LC-600 LC-9A
Catalog No. 5000806549
Encompass
Shimadzu Models LC-600 LC-9A LC-10AD & LC-20AB Self-Priming Outlet Check Valve Assembly Includes Cartridge and Housing OEM No. 228-18522-91
Qty/Pkg: EACH
Catalog No. 50-008-06549 Supplier Analytical Sales & Services Supplier No. 925081
Shimadzu Models LC-600 LC-9A LC-10AD & LC-20AB Self-Priming Outlet Check Valve Assembly Includes Cartridge and Housing OEM No. 228-18522-91
Qty/Pkg: EACH