Learn More
Analytical Sales & Services Shimadzu Models LC-6A & LC-10A
Catalog No. 5000805811
Encompass
Shimadzu Models LC-6A & LC-10AS Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM No. 228-12353-91\n\nQty/Pkg: EACH
Catalog No. 50-008-05811 Supplier Analytical Sales & Services Supplier No. 925182
Shimadzu Models LC-6A & LC-10AS Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM No. 228-12353-91\n\nQty/Pkg: EACH