Learn More
Analytical Sales & Services Shimadzu Models LC-7A LC-10AS
Catalog No. 5000805387
Encompass
Shimadzu Models LC-7A LC-10AS LC-10AT LC-10ATvp LC-2010 & LC-20AT Replacement Teflon Piston Seal OEM No. 228-35145-00 10/PK
Qty/Pkg: 10PK
Catalog No. 50-008-05387 Supplier Analytical Sales & Services Supplier No. 920037
Shimadzu Models LC-7A LC-10AS LC-10AT LC-10ATvp LC-2010 & LC-20AT Replacement Teflon Piston Seal OEM No. 228-35145-00 10/PK
Qty/Pkg: 10PK