Learn More
Analytical Sales & Services Shimadzu Models LC-7A LC-10AS
Supplier: Analytical Sales & Services 925143
Shimadzu Models LC-7A LC-10AS LC-10AT LC-10ATvp LC-2010 & LC-20AT UHMWPE High Density Polyethylene Yellow Piston Seal OEM No. 228-21975-00
Qty/Pkg: EACH