Learn More
Analytical Sales & Services Shimadzu Models LC-7A LC-10AS
Catalog No. 5000804425
Encompass
Shimadzu Models LC-7A LC-10AS LC-10AT LC-10ATvp LC-2010. & LC-20AT UHMW-PE White Piston Wash Seal OEM No. 228-28499-00
Qty/Pkg: EACH
Catalog No. 50-008-04425 Supplier Analytical Sales & Services Supplier No. 925144
Shimadzu Models LC-7A LC-10AS LC-10AT LC-10ATvp LC-2010. & LC-20AT UHMW-PE White Piston Wash Seal OEM No. 228-28499-00
Qty/Pkg: EACH