Learn More
Analytical Sales & Services Shimadzu Outlet Check Valve fo
Catalog No. 5000806194
Encompass
Shimadzu Outlet Check Valve for Models LC-10ATvp & LC-2010 Includes Cartridge and Housing OEM No. 288-34976-91.
Qty/Pkg: EACH
Catalog No. 50-008-06194 Supplier Analytical Sales & Services Supplier No. 925141
Shimadzu Outlet Check Valve for Models LC-10ATvp & LC-2010 Includes Cartridge and Housing OEM No. 288-34976-91.
Qty/Pkg: EACH