Learn More
Analytical Sales & Services Shimadzu Replacement Sapphire
Catalog No. 5000806175
Encompass
Shimadzu Replacement Sapphire Rod Piston for Models LC-10ATvp LC-2010 LC-20AT OEM No. 228-35009-92\n\nQty/Pkg: EACH
Catalog No. 50-008-06175 Supplier Analytical Sales & Services Supplier No. 926010
Shimadzu Replacement Sapphire Rod Piston for Models LC-10ATvp LC-2010 LC-20AT OEM No. 228-35009-92\n\nQty/Pkg: EACH