Learn More
Medchemexpress LLC HY-112066 1mg Medchemexpress, SHIN1 CAS:2146095-85-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1120661MG
Encompass_Preferred
Medchemexpress, HY-112066 1mg SHIN1 CAS:2146095-85-2 SHIN1 (RZ-2994) is a human serine hydroxymethyltransferse 1 and 2 (SHMT1/2) inhibitor with IC50s of 5 and 13 nM, respectively, in an in vitro assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8818