Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-SHIN1 | 2444764-09-2 | 99.4% | 400.47 | 100 MG
SDP

Catalog No. 5000438905
Encompass_Preferred

(-)-SHIN1, also known as (-)-RZ-2994, is an inactive (-) enantiomer of SHIN1. It appears as a solid and is white to off-white in color.

  • Appears as a solid
  • Color is white to off-white
  • Enantiomeric excess of 99.14%
  • Purity (LCMS) of 99.37%

Catalog No. 50-004-38905 Supplier Medchemexpress LLC Supplier No. HY112066B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-SHIN1, also known as (-)-RZ-2994, is an inactive (-) enantiomer of SHIN1. It appears as a solid and is white to off-white in color.

  • Appears as a solid
  • Color is white to off-white
  • Enantiomeric excess of 99.14%
  • Purity (LCMS) of 99.37%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.