Learn More
Medchemexpress LLC (-)-SHIN1 ((-)-RZ-2994) | 2444764-09-2 | 99.1% | 400.47 | 10 MG
SDP
Catalog No. 5000436829
Encompass_Preferred
(-)-SHIN1 ((-)-RZ-2994) is an inactive (-) enantiomer of SHIN1. It is for research use only and not sold to patients.
  • Inactive (-) enantiomer of SHIN1.
  • Available in various quantities as solid or solution (10 mM * 1 mL in DMSO).
  • Purity: 99.14% ee.: 100.00%.
  • For research use only.
  • Can be stored at 4°C under nitrogen (solid) or -80°C/ -20°C in solvent.
Catalog No. 50-004-36829 Supplier Medchemexpress LLC Supplier No. HY112066B10MG
(-)-SHIN1 ((-)-RZ-2994) is an inactive (-) enantiomer of SHIN1. It is for research use only and not sold to patients.
  • Inactive (-) enantiomer of SHIN1.
  • Available in various quantities as solid or solution (10 mM * 1 mL in DMSO).
  • Purity: 99.14% ee.: 100.00%.
  • For research use only.
  • Can be stored at 4°C under nitrogen (solid) or -80°C/ -20°C in solvent.