Learn More
Medchemexpress LLC HY-112700 5mg Medchemexpress, SHIP2-IN-1 CAS:2252247-80-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127005MG
Encompass_Preferred
Medchemexpress, HY-112700 5mg SHIP2-IN-1 CAS:2252247-80-4 SHIP2-IN-1 is a potent SHIP2 inhibitor, inhibits SHIP2 activity, with an IC50 of 2 µM. SHIP2-IN-1 blocks GSK3β activation by phosphorylation at the Ser9 residue. SHIP2-IN-1 is used in the research of Alzheimer’s disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0825