Learn More
Echelon Biosciences Research Labs 5 PtdIns(3,4,5)P3 Phosphatase Activity Fluorescence Polarization Assay
SDP
Catalog No. 50720425
Encompass
The 5 PtdIns(3,4,5)P3 Phosphatase Activity Fluorescence Polarization Assay determines activity through quantification of the end product, PI(3,4)P2.Sample Type: in vitro determination of 5 phosphatase activity in cancer tissues and cell lines and compound screening for drug discoverySample Volume: 2.5-5.0 L enzyme / inhibitorAssay Range: 0.0625 M to 2.0 M This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-425 Supplier Echelon Biosciences Research Labs Supplier No. K1400EAKIT
The 5 PtdIns(3,4,5)P3 Phosphatase Activity Fluorescence Polarization Assay determines activity through quantification of the end product, PI(3,4)P2.Sample Type: in vitro determination of 5 phosphatase activity in cancer tissues and cell lines and compound screening for drug discoverySample Volume: 2.5-5.0 L enzyme / inhibitorAssay Range: 0.0625 M to 2.0 M This item is non returnable