Learn More
Berlin Packaging Hazardous Material Tall Paint Can Shipping System, 32 oz Narrow Mouth Bottle, 4/cs
Catalog No. NC1677309
Encompass_Preferred
This shipping system ships inner containers of 32 oz (1 L) or less, secondary inner packaging: slip cover can. Ideal for ground shipments. Dangerous Goods’ Special Permit Packaging, DOT-SP 9168, can be used to ship Packing Groups I, II, or III materials by all modes of transport.
Catalog No. NC1677309 Supplier Berlin Packaging Supplier No. HMS66421M
This shipping system ships inner containers of 32 oz (1 L) or less, secondary inner packaging: slip cover can. Ideal for ground shipments. Dangerous Goods’ Special Permit Packaging, DOT-SP 9168, can be used to ship Packing Groups I, II, or III materials by all modes of transport.