Learn More
Mitegen Upright Shipping Case for the IC Biomedical CX100, CXR100, 4D, or 4DX Dryshippers, Provides High-Impact Protection, Has Domed Top With Wide Base
SDP
Catalog No. NC2292014
Encompass
Upright Shipping Case delivers superior protection for nitrogen vapor dryshippers during rigorous handling and extended transit times. Constructed from high-impact-resistant material, this robust case safeguards its valuable contents from physical damage, ensuring sample integrity throughout the journey. Its unique mushroom-designed domed top effectively prevents stacking, while the wide circular base encourages stable upright orientation, which is crucial for maintaining optimal sample conditions. For enhanced security, a built-in slot allows the case to be easily locked down. This Mitegen shipping case is specifically engineered to accommodate various Mitegen Cryo-Express dryshippers, including models such as the CX100, CXR100, 4D, or 4DX, providing a safer, more secure enclosure for your cryogenic samples. Key specifications include a height of 24 inches, a base diameter of 15 inches, and a weight of 14 lbs. Lockable, 24"H x 15"D, 14 lbs
This item is not returnable. View return policy
Catalog No. NC2292014 Supplier Mitegen Supplier No. TW4LSC
Upright Shipping Case delivers superior protection for nitrogen vapor dryshippers during rigorous handling and extended transit times. Constructed from high-impact-resistant material, this robust case safeguards its valuable contents from physical damage, ensuring sample integrity throughout the journey. Its unique mushroom-designed domed top effectively prevents stacking, while the wide circular base encourages stable upright orientation, which is crucial for maintaining optimal sample conditions. For enhanced security, a built-in slot allows the case to be easily locked down. This Mitegen shipping case is specifically engineered to accommodate various Mitegen Cryo-Express dryshippers, including models such as the CX100, CXR100, 4D, or 4DX, providing a safer, more secure enclosure for your cryogenic samples. Key specifications include a height of 24 inches, a base diameter of 15 inches, and a weight of 14 lbs. Lockable, 24"H x 15"D, 14 lbs