Learn More
MSC Packaging Wholesalers Box, 9.5 x 9.5 x 7, 1 ea
Catalog No. 501920598
Encompass
This insulated shipping container is designed to maintain the temperature integrity of sensitive products during transport. Its durable construction protects contents from damage while reducing shipping costs.
  • Maintains insulation and integrity of temperature-sensitive products
  • Lightweight, reusable, and recyclable construction
  • Cartons printed with Rush, Perishable, and This Side Up shipping instructions
  • Protects contents from damage and maintains ideal temperatures
  • Features 1.25 in thick EPS foam body with 32 ECT corrugated carton
  • Made in America
Catalog No. 50-192-0598 Supplier MSC Supplier No. 39559273
This insulated shipping container is designed to maintain the temperature integrity of sensitive products during transport. Its durable construction protects contents from damage while reducing shipping costs.
  • Maintains insulation and integrity of temperature-sensitive products
  • Lightweight, reusable, and recyclable construction
  • Cartons printed with Rush, Perishable, and This Side Up shipping instructions
  • Protects contents from damage and maintains ideal temperatures
  • Features 1.25 in thick EPS foam body with 32 ECT corrugated carton
  • Made in America