Learn More
BINDER™ Shipping Pallet
SDP
Catalog No. 502875742 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB260, KB470, KB720, KB1060
KBF260, KBF470, KBF720, KBF1060
KBFPRO260, KBFPRO470, KBFPRO720
KBPRO260, KBPRO470, KBPRO720
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5742 KB260, KB470, KB720, KB1060
50-287-5675 KBPRO260, KBPRO470, KBPRO720
50-287-5613 KBF260, KBF470, KBF720, KBF1060
50-287-5625 KBFPRO260, KBFPRO470, KBFPRO720
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-287-5742 Supplier BINDER™ Supplier No. 80122545
Add to Cart
Add to Cart
Shipping pallet with unloading ramp for installing unit without forklift
For
Shipping pallet with unloading ramp for easy unloading of chamber from shipping pallet.
Specifications
For Use With (Equipment) KB260, KB470, KB720, KB1060
Type Platform
Material Fiber