Learn More
Saf T Pak Inc STT-410-500 - Saf-T-Temp -10°C Refrigerated PCM Freezer Paks, 500g, 16/Carton
SDP
Catalog No. 501904618 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
Saf-T-Pak's Phase Change Material (Saf-T-Temp) is designed to provide your shipment with the thermal protection needed to ensure safe transport.SAF-T-Temp Freezer Pak (-10°C) Phase Change Material. 500 Gram PCM Pak. This is a smaller Phase Change Material Pak, designed to keep a frozen environment throughout transit.Shipping specimens needing to remain frozenOur PCM provides a perfect thermal barrier/insulation between your payload and external temperature fluctuations frequently seen during transport.FEATURES: 500 gram Paks16 per carton
Catalog No. 50-190-4618 Supplier Saf T Pak Inc Supplier No. 35144
Saf-T-Pak's Phase Change Material (Saf-T-Temp) is designed to provide your shipment with the thermal protection needed to ensure safe transport.SAF-T-Temp Freezer Pak (-10°C) Phase Change Material. 500 Gram PCM Pak. This is a smaller Phase Change Material Pak, designed to keep a frozen environment throughout transit.Shipping specimens needing to remain frozenOur PCM provides a perfect thermal barrier/insulation between your payload and external temperature fluctuations frequently seen during transport.FEATURES: 500 gram Paks16 per carton